2-Amino-5-methoxyphenol
Lieferant: Apollo Scientific
|
Achtung
|
OR110697-1G
OR110697-250MG
APOSOR110697-1GEA
0
EUR
APOSOR110697-1G
APOSOR110697-250MG
2-Amino-5-methoxyphenol
2-Amino-5-methoxyphenol
Formel:
C₇H₉NO₂ Schmelzpunkt: 126…129 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD03840435 CAS-Nummer: 40925-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...