6-Chloro-N4-methylpyrimidine-2,4-diamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-chloro-6-(methylamino)pyrimidine, 2-Amino-4-chloro-6-(methylamino)-1,3-diazine

OR110703-1G OR110703-10G OR110703-5G
APOSOR110703-1GEA 63.8 EUR
APOSOR110703-1G APOSOR110703-10G APOSOR110703-5G
6-Chloro-N4-methylpyrimidine-2,4-diamine
6-Chloro-N4-methylpyrimidine-2,4-diamine
Formel: C₅H₇ClN₄
Molecular Weight: 158,589 g/mol
Schmelzpunkt: 159…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091108
CAS-Nummer: 1005-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR