2-Chloro-N-(2,4-dimethoxybenzyl)acetamide

Lieferant: Apollo Scientific
Achtung

OR110742-1G OR110742-250MG
APOSOR110742-1GEA 126 EUR
APOSOR110742-1G APOSOR110742-250MG
2-Chloro-N-(2,4-dimethoxybenzyl)acetamide
2-Chloro-N-(2,4-dimethoxybenzyl)acetamide
Formel: C₁₁H₁₄ClNO₃
Molecular Weight: 243,687 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09673384
CAS-Nummer: 928713-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR