Methyl 4-bromo-1H-benzimidazole-6-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-6-(methoxycarbonyl)-1H-benzimidazole

OR110783-1G OR110783-250MG
APOSOR110783-1GEA 322 EUR
APOSOR110783-1G APOSOR110783-250MG
Methyl 4-bromo-1H-benzimidazole-6-carboxylate
Methyl 4-bromo-1H-benzimidazole-6-carboxylate
Formel: C₉H₇BrN₂O₂
Schmelzpunkt: 256…259 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22421935
CAS-Nummer: 1354756-19-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR