(3,3-Dimethoxy-1-methylpropylidene)malononitrile

Lieferant: Apollo Scientific
Gefahr

OR110842-1G OR110842-250MG
APOSOR110842-1GEA 998 EUR
APOSOR110842-1G APOSOR110842-250MG
(3,3-Dimethoxy-1-methylpropylidene)malononitrile
(3,3-Dimethoxy-1-methylpropylidene)malononitrile
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180,204 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09030793
CAS-Nummer: 410547-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR