8-Bromo-6-chlorocoumarin-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Bromo-3-carboxy-6-chlorocoumarin, 8-Bromo-3-carboxy-6-chloro-2-oxo-2H-1-benzopyran, 8-Bromo-6-chloro-2-oxo-2H-chromene-3-carboxylic acid
OR110849-1G
OR110849-5G
APOSOR110849-1GEA
157
EUR
APOSOR110849-1G
APOSOR110849-5G
8-Bromo-6-chlorocoumarin-3-carboxylic acid
8-Bromo-6-chlorocoumarin-3-carboxylic acid
Formel:
C₁₀H₄BrClO₄ Molecular Weight: 303,493 g/mol Schmelzpunkt: 236…238 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03406937 CAS-Nummer: 213749-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...