5-Bromo-N-methoxy-N-methyl-2-furamide

Lieferant: Apollo Scientific
Achtung

OR110852-1G OR110852-500MG
APOSOR110852-1GEA 364 EUR
APOSOR110852-1G APOSOR110852-500MG
5-Bromo-N-methoxy-N-methyl-2-furamide
5-Bromo-N-methoxy-N-methyl-2-furamide
Formel: C₇H₈BrNO₃
Molecular Weight: 234,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16104416
CAS-Nummer: 179055-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR