(1-Oxoisoindolin-2-yl)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1-Oxoisoindolin-2-yl)acetic acid, (1,3-Dihydro-1-oxo-2H-isoindol-2-yl)acetic acid, 2-(Carboxymethyl)-2,3-dihydro-1-oxo-1H-isoindole
OR110884-10G
OR110884-1G
OR110884-5G
APOSOR110884-10GEA
369
EUR
APOSOR110884-10G
APOSOR110884-1G
APOSOR110884-5G
(1-Oxoisoindolin-2-yl)acetic acid
(1-Oxoisoindolin-2-yl)acetic acid
Formel:
C₁₀H₉NO₃ Schmelzpunkt: 196…199 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00664645 CAS-Nummer: 39221-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...