3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR110886-1G
OR110886-5G
APOSOR110886-1GEA
116
EUR
APOSOR110886-1G
APOSOR110886-5G
3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
Formel:
C₁₅H₁₁NO₃ Molecular Weight: 253,253 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04358395 CAS-Nummer: 4770-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...