Ethyl 4-[(4-chlorobutanoyl)amino]benzoate

Lieferant: Apollo Scientific

OR110899-1G OR110899-5G
APOSOR110899-1GEA 92.8 EUR
APOSOR110899-1G APOSOR110899-5G
Ethyl 4-[(4-chlorobutanoyl)amino]benzoate
Ethyl 4-[(4-chlorobutanoyl)amino]benzoate
Formel: C₁₃H₁₆ClNO₃
Molecular Weight: 269,724 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00204045
CAS-Nummer: 139348-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR