1-(5-Chloro-1-benzofuran-2-yl)ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Chlorobenzofuran-2-yl)ethanone

OR110910-1G OR110910-250MG
APOSOR110910-1GEA 174 EUR
APOSOR110910-1G APOSOR110910-250MG
1-(5-Chloro-1-benzofuran-2-yl)ethanone
1-(5-Chloro-1-benzofuran-2-yl)ethanone
Formel: C₁₀H₇ClO₂
Molecular Weight: 194.614 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00661909
CAS-Nummer: 1646-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR