6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR110920-1G OR110920-5G
APOSOR110920-1GEA 371 EUR
APOSOR110920-1G APOSOR110920-5G
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carboxylic acid
6,7-Dihydro-4H-thieno[3,2-c]pyran-2-carboxylic acid
Formel: C₈H₈O₃S
Molecular Weight: 184,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12088019
CAS-Nummer: 933747-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR