5-Bromo-4-methyl-7-nitro-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR110944-1G OR110944-5G
APOSOR110944-1GEA 69.6 EUR
APOSOR110944-1G APOSOR110944-5G
5-Bromo-4-methyl-7-nitro-1H-indazole
5-Bromo-4-methyl-7-nitro-1H-indazole
Formel: C₈H₆BrN₃O₂
Molecular Weight: 256.056 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26939176
CAS-Nummer: 1858255-52-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR