Ethyl (3-amino-4-hydroxyphenyl)acetate

Lieferant: Apollo Scientific
Achtung

OR110954-1G OR110954-250MG
APOSOR110954-1GEA 662 EUR
APOSOR110954-1G APOSOR110954-250MG
Ethyl (3-amino-4-hydroxyphenyl)acetate
Ethyl (3-amino-4-hydroxyphenyl)acetate
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00230584
CAS-Nummer: 86818-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR