N-[4-(4-Bromophenoxy)benzyl]-N-methylamine

Lieferant: Apollo Scientific

OR110981-1G OR110981-5G
APOSOR110981-1GEA 198 EUR
APOSOR110981-1G APOSOR110981-5G
N-[4-(4-Bromophenoxy)benzyl]-N-methylamine
N-[4-(4-Bromophenoxy)benzyl]-N-methylamine
Formel: C₁₄H₁₄BrNO
Molecular Weight: 292,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11607780
CAS-Nummer: 1082875-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR