1-(3-Nitrophenyl)pyrrolidine

Lieferant: Apollo Scientific

OR110999-1G OR110999-10G OR110999-5G
APOSOR110999-1GEA 0 EUR
APOSOR110999-1G APOSOR110999-10G APOSOR110999-5G
1-(3-Nitrophenyl)pyrrolidine
1-(3-Nitrophenyl)pyrrolidine
Formel: C₁₀H₁₂N₂O₂
Molecular Weight: 192,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02940746
CAS-Nummer: 132993-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR