Ethyl thiophene-2-carboximidoate hydrochloride
Lieferant: Apollo Scientific
OR111000-1G OR111000-5G
APOSOR111000-1GEA 0 EUR
APOSOR111000-1G APOSOR111000-5G
Ethyl thiophene-2-carboximidoate hydrochloride
Ethyl thiophene-2-carboximidoate hydrochloride
Formel: C₇H₉NOS·ClH
Molecular Weight: 191.678 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09953263
CAS-Nummer: 54610-52-5