1-Bromo-4-(isopropylthio)benzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-(propan-2-ylsulphanyl)benzene, (4-Bromophenyl)(isopropyl)sulphane

OR111003-1G OR111003-100MG OR111003-5G
APOSOR111003-1GEA 132 EUR
APOSOR111003-1G APOSOR111003-100MG APOSOR111003-5G
1-Bromo-4-(isopropylthio)benzene
1-Bromo-4-(isopropylthio)benzene
Formel: C₉H₁₁BrS
Molecular Weight: 231.153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20484423
CAS-Nummer: 70398-89-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR