1-[2-(Benzyloxy)-5-methylphenyl]ethanone
Lieferant: Apollo Scientific
Synonyme:
1-(2-(Benzyloxy)-5-methylphenyl)ethanone
OR111018-1G
OR111018-5G
APOSOR111018-1GEA
198
EUR
APOSOR111018-1G
APOSOR111018-5G
1-[2-(Benzyloxy)-5-methylphenyl]ethanone
1-[2-(Benzyloxy)-5-methylphenyl]ethanone
Formel:
C₁₆H₁₆O₂ Molecular Weight: 240,297 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12638339 CAS-Nummer: 36808-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...