3-(Piperidin-1-ylmethyl)benzonitrile

Lieferant: Apollo Scientific

OR111032-1G OR111032-5G
APOSOR111032-1GEA 0 EUR
APOSOR111032-1G APOSOR111032-5G
3-(Piperidin-1-ylmethyl)benzonitrile
3-(Piperidin-1-ylmethyl)benzonitrile
Formel: C₁₃H₁₆N₂
Molecular Weight: 200,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09927536
CAS-Nummer: 857284-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR