1-(3-Bromophenyl)-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR111055-1G OR111055-5G
APOSOR111055-1GEA 0 EUR
APOSOR111055-1G APOSOR111055-5G
1-(3-Bromophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Bromophenyl)-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₁H₁₀BrNO₃
Molecular Weight: 284,106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06001923
CAS-Nummer: 714207-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR