2-Bromo-1-(2,3-dihydro-1-benzofuran-5-yl)ethanone

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(2,3-dihydrobenzofuran-5-yl)ethanone

OR111097-1G OR111097-250MG
APOSOR111097-1GEA 256 EUR
APOSOR111097-1G APOSOR111097-250MG
2-Bromo-1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
2-Bromo-1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
Formel: C₁₀H₉BrO₂
Molecular Weight: 241,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08444468
CAS-Nummer: 151427-19-9
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR