6,8-Dibromo-5-chloroquinazolin-4-ol

Lieferant: Apollo Scientific

OR111102-1G OR111102-250MG
APOSOR111102-1GEA 232 EUR
APOSOR111102-1G APOSOR111102-250MG
6,8-Dibromo-5-chloroquinazolin-4-ol
6,8-Dibromo-5-chloroquinazolin-4-ol
Formel: C₈H₃Br₂ClN₂O
Molecular Weight: 338,383 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954638
CAS-Nummer: 1858241-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR