1-(4-Nitrobenzoyl)piperidine

Lieferant: Apollo Scientific

OR111114-1G OR111114-5G
APOSOR111114-1GEA 0 EUR
APOSOR111114-1G APOSOR111114-5G
1-(4-Nitrobenzoyl)piperidine
1-(4-Nitrobenzoyl)piperidine
Formel: C₁₂H₁₄N₂O₃
Molecular Weight: 234,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00444400
CAS-Nummer: 20857-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR