2-Methylcyclopropane-1-carbohydrazide
Lieferant: Apollo Scientific
OR111120-1G
OR111120-5G
OR111120-10G
APOSOR111120-1GEA
223
EUR
APOSOR111120-1G
APOSOR111120-5G
APOSOR111120-10G
2-Methylcyclopropane-1-carbohydrazide
2-Methylcyclopropane-1-carbohydrazide
Formel:
C₅H₁₀N₂O Molecular Weight: 114,146 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01722433 CAS-Nummer: 63884-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...