4-Imidazo[1,2-a]pyrimidin-2-ylbenzonitrile

Lieferant: Apollo Scientific

OR111170-10G OR111170-1G OR111170-5G
APOSOR111170-10GEA 0 EUR
APOSOR111170-10G APOSOR111170-1G APOSOR111170-5G
4-Imidazo[1,2-a]pyrimidin-2-ylbenzonitrile
4-Imidazo[1,2-a]pyrimidin-2-ylbenzonitrile
Formel: C₁₃H₈N₄
Molecular Weight: 220.229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02680815
CAS-Nummer: 104691-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR