[(3-Chlorophenyl)thio]acetonitrile

Lieferant: Apollo Scientific

OR111217-10G OR111217-5G
APOSOR111217-10GEA 0 EUR
APOSOR111217-10G APOSOR111217-5G
[(3-Chlorophenyl)thio]acetonitrile
[(3-Chlorophenyl)thio]acetonitrile
Formel: C₈H₆ClNS
Molecular Weight: 183,658 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16835766
CAS-Nummer: 18527-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR