Ethyl 5-chloro-2-hydroxynicotinate

Lieferant: Apollo Scientific
Achtung

OR111283-1G OR111283-5G
APOSOR111283-1GEA 319 EUR
APOSOR111283-1G APOSOR111283-5G
Ethyl 5-chloro-2-hydroxynicotinate
Ethyl 5-chloro-2-hydroxynicotinate
Formel: C₈H₈ClNO₃
Molecular Weight: 201,607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698193
CAS-Nummer: 1214366-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR