Sodium (1Z)-2-(dimethoxymethyl)-3-methoxy-3-oxoprop-1-en-1-olate

Lieferant: Apollo Scientific
Achtung

OR111383-1G
APOSOR111383-1GEA 618 EUR
APOSOR111383-1G
Sodium (1Z)-2-(dimethoxymethyl)-3-methoxy-3-oxoprop-1-en-1-olate
Sodium (1Z)-2-(dimethoxymethyl)-3-methoxy-3-oxoprop-1-en-1-olate
Formel: C₇H₁₁O₅·Na
Molecular Weight: 198,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347677
CAS-Nummer: 927806-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR