7-Bromo-1-benzothiophene-2-carbonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR111393-1G OR111393-250MG
APOSOR111393-1GEA 375 EUR
APOSOR111393-1G APOSOR111393-250MG
7-Bromo-1-benzothiophene-2-carbonyl chloride
7-Bromo-1-benzothiophene-2-carbonyl chloride
Formel: C₉H₄BrClOS
Molecular Weight: 275,549 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28992243
CAS-Nummer: 1935390-73-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR