3-Amino-3-(2,3-dihydro-1,4-benzodioxin-6-yl)propanoic acid

Lieferant: Apollo Scientific
Achtung

OR111399-1G OR111399-250MG
APOSOR111399-1GEA 232 EUR
APOSOR111399-1G APOSOR111399-250MG
3-Amino-3-(2,3-dihydro-1,4-benzodioxin-6-yl)propanoic acid
3-Amino-3-(2,3-dihydro-1,4-benzodioxin-6-yl)propanoic acid
Formel: C₁₁H₁₃NO₄
Molecular Weight: 223.225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871320
CAS-Nummer: 138621-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR