2-Bromo-4-methylbenzyl chloride

Lieferant: Apollo Scientific
Gefahr

OR111406-1G OR111406-250MG
APOSOR111406-1GEA 0 EUR
APOSOR111406-1G APOSOR111406-250MG
2-Bromo-4-methylbenzyl chloride
2-Bromo-4-methylbenzyl chloride
Formel: C₈H₈BrCl
Lagertemperatur: Raumtemperatur
MDL: MFCD12025346
CAS-Nummer: 147542-02-7
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR