(4-Amino-3-bromophenyl)acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-3-bromophenylacetic acid

OR111430-1G OR111430-5G
APOSOR111430-1GEA 114 EUR
APOSOR111430-1G APOSOR111430-5G
(4-Amino-3-bromophenyl)acetic acid
(4-Amino-3-bromophenyl)acetic acid
Formel: C₈H₈BrNO₂
Molecular Weight: 230,059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09842457
CAS-Nummer: 66955-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR