2-Chloro-1-(chloromethyl)-3,4-dimethoxybenzene
Lieferant: Apollo Scientific
OR111435-1G
OR111435-5G
OR111435-250MG
APOSOR111435-1GEA
155
EUR
APOSOR111435-1G
APOSOR111435-5G
APOSOR111435-250MG
2-Chloro-1-(chloromethyl)-3,4-dimethoxybenzene
2-Chloro-1-(chloromethyl)-3,4-dimethoxybenzene
Formel:
C₉H₁₀Cl₂O₂ Molecular Weight: 221.08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18850385 CAS-Nummer: 93983-14-3 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...