2-Chloro-1-propyl-1H-indole-3-carbaldehyde

Lieferant: Apollo Scientific

OR111463-1G OR111463-5G
APOSOR111463-1GEA 0 EUR
APOSOR111463-1G APOSOR111463-5G
2-Chloro-1-propyl-1H-indole-3-carbaldehyde
2-Chloro-1-propyl-1H-indole-3-carbaldehyde
Formel: C₁₂H₁₂ClNO
Molecular Weight: 221,683 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19309611
CAS-Nummer: 64788-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR