N-(4-Chlorophenyl)-4-nitrobenzamide

Lieferant: Apollo Scientific

OR111490-1G OR111490-5G
APOSOR111490-1GEA 0 EUR
APOSOR111490-1G APOSOR111490-5G
N-(4-Chlorophenyl)-4-nitrobenzamide
N-(4-Chlorophenyl)-4-nitrobenzamide
Formel: C₁₃H₉ClN₂O₃
Molecular Weight: 276,675 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00430502
CAS-Nummer: 2585-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR