2,4-Dichloro-N-methylbenzenesulphonamide
Lieferant: Apollo Scientific
|
Achtung
|
OR111507-1G
OR111507-5G
APOSOR111507-1GEA
79.9
EUR
APOSOR111507-1G
APOSOR111507-5G
2,4-Dichloro-N-methylbenzenesulphonamide
2,4-Dichloro-N-methylbenzenesulphonamide
Formel:
C₇H₇Cl₂NO₂S Molecular Weight: 240,107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03989572 CAS-Nummer: 565181-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...