2,4-Dichloro-3,5-dimethylphenyl acetate

Lieferant: Apollo Scientific

OR111602-1G OR111602-5G
APOSOR111602-1GEA 248 EUR
APOSOR111602-1G APOSOR111602-5G
2,4-Dichloro-3,5-dimethylphenyl acetate
2,4-Dichloro-3,5-dimethylphenyl acetate
Formel: C₁₀H₁₀Cl₂O₂
Molecular Weight: 233,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08273809
CAS-Nummer: 99421-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR