2,3-Dimethyl-6,7-dimethoxyquinoxaline 98%

Lieferant: Apollo Scientific

Synonyme: 6,7-Dimethoxy-2,3-dimethylquinoxaline

OR11171-100MG OR11171-1G OR11171-250MG
APOSOR11171-100MGEA 79.9 EUR
APOSOR11171-100MG APOSOR11171-1G APOSOR11171-250MG
2,3-Dimethyl-6,7-dimethoxyquinoxaline 98%
2,3-Dimethyl-6,7-dimethoxyquinoxaline
Formel: C₁₂H₁₄N₂O₂
Molecular Weight: 218,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038946
CAS-Nummer: 32388-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR