4-Amino-3',5'-dichlorobiphenyl

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(3,5-Dichlorophenyl)aniline, 3',5'-Dichloro-[1,1'-biphenyl]-4-amine

OR11339-25G OR11339-5G OR11339-100G
APOSOR11339-25GEA 290 EUR
APOSOR11339-25G APOSOR11339-5G APOSOR11339-100G
4-Amino-3',5'-dichlorobiphenyl
4-Amino-3',5'-dichlorobiphenyl
Formel: C₁₂H₉Cl₂N
Molecular Weight: 238,113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06796293
CAS-Nummer: 405058-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR