4-Chloro-6-(2-methyl-1H-imidazol-1-yl)pyrimidine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(6-Chloropyrimidin-4-yl)-2-methyl-1H-imidazole

OR11462-1G OR11462-5G
APOSOR11462-1GEA 151 EUR
APOSOR11462-1G APOSOR11462-5G
4-Chloro-6-(2-methyl-1H-imidazol-1-yl)pyrimidine 98%
4-Chloro-6-(2-methyl-1H-imidazol-1-yl)pyrimidine
Formel: C₈H₇ClN₄
Molecular Weight: 194.621 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09475881
CAS-Nummer: 941294-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR