4-Chloro-6-(4-methyl-1H-imidazol-1-yl)pyrimidine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(6-Chloropyrimidin-4-yl)-4-methyl-1H-imidazole

OR11463-1G OR11463-5G OR11463-25G
APOSOR11463-1GEA 151 EUR
APOSOR11463-1G APOSOR11463-5G APOSOR11463-25G
4-Chloro-6-(4-methyl-1H-imidazol-1-yl)pyrimidine 98%
4-Chloro-6-(4-methyl-1H-imidazol-1-yl)pyrimidine
Formel: C₈H₇ClN₄
Molecular Weight: 194,621 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09475883
CAS-Nummer: 941294-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR