5-Bromo-2-methoxynicotinonitrile 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Bromo-2-methoxypyridine-3-carbonitrile, 5-Bromo-3-cyano-2-methoxypyridine
OR11516-1G
OR11516-5G
APOSOR11516-1GEA
79.9
EUR
APOSOR11516-1G
APOSOR11516-5G
5-Bromo-2-methoxynicotinonitrile 98%
5-Bromo-2-methoxynicotinonitrile
Formel:
C₇H₅BrN₂O Molecular Weight: 213.031 g/mol Schmelzpunkt: 110…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09801047 CAS-Nummer: 941294-54-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...