3-Bromo-5-(methylsulphonyl)pyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-methylsulphonylpyridine, 5-Bromopyridin-3-yl methyl sulphone

OR11568-1G OR11568-5G
APOSOR11568-1GEA 72.5 EUR
APOSOR11568-1G APOSOR11568-5G
3-Bromo-5-(methylsulphonyl)pyridine 95%
3-Bromo-5-(methylsulphonyl)pyridine
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801057
CAS-Nummer: 445491-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR