2,7-Dibromo-2',3',5',6'-tetrahydrospiro[fluorene-9,4'-pyran]

Lieferant: Apollo Scientific
Achtung

OR11597-1G
APOSOR11597-1GEA 63.8 EUR
APOSOR11597-1G
2,7-Dibromo-2',3',5',6'-tetrahydrospiro[fluorene-9,4'-pyran]
2,7-Dibromo-2',3',5',6'-tetrahydrospiro[fluorene-9,4'-pyran]
Formel: C₁₇H₁₄Br₂O
Molecular Weight: 394,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800926
CAS-Nummer: 934269-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR