1H-Pyrrole-2-carboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Carbamoyl-1H-pyrrole, 2-(Aminocarbonyl)-1H-pyrrole

OR11616-1G OR11616-5G
APOSOR11616-1GEA 179 EUR
APOSOR11616-1G APOSOR11616-5G
1H-Pyrrole-2-carboxamide
1H-Pyrrole-2-carboxamide
Formel: C₅H₆N₂O
Molecular Weight: 110,114 g/mol
Schmelzpunkt: 173…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00273121
CAS-Nummer: 4551-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR