4-(4-Bromobenzoyl)morpholine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Bromophenyl)(morpholin-4-yl)methanone, (4-Bromophenyl)(morpholino)methanone
OR11622-5G
OR11622-1G
APOSOR11622-5GEA
127
EUR
APOSOR11622-5G
APOSOR11622-1G
4-(4-Bromobenzoyl)morpholine
4-(4-Bromobenzoyl)morpholine
Formel:
C₁₁H₁₂BrNO₂ Molecular Weight: 270,122 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00447938 CAS-Nummer: 127580-92-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...