4-(3-Bromobenzoyl)morpholine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Bromophenyl)(morpholin-4-yl))methanone
OR11927-1G
OR11927-5G
OR11927-25G
APOSOR11927-1GEA
88.6
EUR
APOSOR11927-1G
APOSOR11927-5G
APOSOR11927-25G
4-(3-Bromobenzoyl)morpholine
4-(3-Bromobenzoyl)morpholine
Formel:
C₁₁H₁₂BrNO₂ Molecular Weight: 270,122 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00194119 CAS-Nummer: 153435-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...