4-Methoxyphenyl methylsulphonate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methoxyphenyl mesylate, 4-[(Methylsulphonyl)oxy]anisole

OR11951-25G OR11951-5G
APOSOR11951-25GEA 789 EUR
APOSOR11951-25G APOSOR11951-5G
4-Methoxyphenyl methylsulphonate 98%
4-Methoxyphenyl methylsulphonate
Formel: C₈H₁₀O₄S
Molecular Weight: 202.228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01231360
CAS-Nummer: 19013-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR