3-(Pyrimidin-2-yl)benzyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-[3-(Chloromethyl)phenyl]pyrimidine, 2-(3-(Chloromethyl)phenyl)pyrimidine
OR12242-1G
OR12242-250MG
APOSOR12242-1GEA
468
EUR
APOSOR12242-1G
APOSOR12242-250MG
3-(Pyrimidin-2-yl)benzyl chloride 97%
3-(Pyrimidin-2-yl)benzyl chloride
Formel:
C₁₁H₉ClN₂ Molecular Weight: 204,656 g/mol Schmelzpunkt: 68…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09064978 CAS-Nummer: 898289-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...